APO447397830 Apollo Scientific Ltd

(2,2-Difluoroethoxy)benzene

  • MDL number MFCD19442073
  • Cas Number : 1309602-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72583

Safety Information

signalword Warning
pcodes P210;P280;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72583
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart