APO447397852 Apollo Scientific Ltd

2,2,3,3,4,4,5,5,5-Nonafluoropentyl difluoromethylether

  • MDL number MFCD27918475
  • Cas Number : 1661005-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Physical Form Liquid
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72605

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72605
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart