APO447397861 Apollo Scientific Ltd

3-(2-Chloroethoxy)-1,1,2,2-tetrafluoropropane

  • MDL number MFCD11164793
  • Cas Number : 64769-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72614

Safety Information

signalword Warning
pcodes P210;P280;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72614
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart