APO447397863 Apollo Scientific Ltd

5-Chloro-1,1-difluoro-3-oxapentane

  • MDL number MFCD19442046
  • Cas Number : 1309602-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72616

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=72616
Availability details for
Quantity
  • Availability
  • Add to Cart