APO447397904 Apollo Scientific Ltd

Ethyl (2R)-2-fluoropropanoate

  • MDL number MFCD28054290
  • Cas Number : 72959-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 93-94/300mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72657

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72657
pictograms GHS02;GHS07
hstatements H226;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart