APO447397917 Apollo Scientific Ltd

2,2-Difluorobutane

  • MDL number MFCD20489351
  • Cas Number : 353-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -114
Purity 97
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 30-31
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72670

Safety Information

signalword Danger
pcodes P210;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72670
pictograms GHS02;GHS07;GHS04
hstatements H225;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart