APO448688165 Apollo Scientific Ltd

4-Amino-3-bromo-5-fluorobenzotrifluoride

  • MDL number MFCD28397844
  • Cas Number : 1034325-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72926

Safety Information

signalword Danger
pcodes P280;P301+P310;P302+P352
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72926
pictograms GHS06
hstatements H301;H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart