APO448688167 Apollo Scientific Ltd

2-Bromo-3-fluoro-4-iodoaniline

  • MDL number MFCD28397846
  • Cas Number : 1936364-85-0
  • Molecular Weight : 315.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-fluoro-4-iodoaniline

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart