APO448688203 Apollo Scientific Ltd

4-Bromo-N,3-dimethyl-N-methoxybenzamide

  • MDL number MFCD26802226
  • Cas Number : 170230-01-0
  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-n3-dimethyl-n-methoxybenzamide

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-n3-dimethyl-n-methoxybenzamide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart