APO448688243 Apollo Scientific Ltd

2-Bromo-3-nitroaniline

  • MDL number MFCD12923157
  • Cas Number : 35757-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-nitroaniline

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-nitroaniline
pictograms GHS06;GHS08
hstatements H301+H311+H331;H373
Availability details for
Quantity
  • Availability
  • Add to Cart