APO448688250 Apollo Scientific Ltd

4-Iodo-3-methoxybenzaldehyde

  • MDL number MFCD11847312
  • Cas Number : 121404-83-9
  • Molecular Weight : 262.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-iodo-3-methoxybenzaldehyde
Inchi Code 1S/C8H7IO2/c1-11-8-4-6(5-10)2-3-7(8)9/h2-5H,1H3
Inchi Key PHSPBXTVMJOTQN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-iodo-3-methoxybenzaldehyde

Safety Information

signalword Danger
pcodes P280;P301+P310
MSDS https://store.apolloscientific.co.uk/product/4-iodo-3-methoxybenzaldehyde
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart