APO448688279 Apollo Scientific Ltd

2-Bromo-6-chloroiodobenzene

  • MDL number MFCD09907935
  • Cas Number : 450412-28-9
  • Molecular Weight : 317.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-3-chloro-2-iodobenzene
Inchi Code 1S/C6H3BrClI/c7-4-2-1-3-5(8)6(4)9/h1-3H
Inchi Key GIMVCZMZRZGDTL-UHFFFAOYSA-N

Description

Synonym 1-Bromo-3-chloro-2-iodobenzene
Purity 99%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-chloroiodobenzene

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-chloroiodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart