APO448688287 Apollo Scientific Ltd

4-Bromo-3-methylbenzyl bromide

  • MDL number MFCD09909629
  • Cas Number : 27561-51-9
  • Molecular Weight : 263.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-methylbenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-methylbenzyl-bromide
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart