APO448688364 Apollo Scientific Ltd

3-Fluoro-2-nitrobenzotrifluoride

  • MDL number MFCD12032180
  • Cas Number : 1214335-98-4
  • Molecular Weight : 209.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-2-nitrobenzotrifluoride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-2-nitrobenzotrifluoride
pictograms GHS06;GHS07
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart