APO448899927 Apollo Scientific Ltd

4-Amino-3,5-difluorobenzoic acid

  • MDL number MFCD03407440
  • Cas Number : 500577-99-1
  • Molecular Weight : 173.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-3,5-difluorobenzoic acid
Inchi Code 1S/C7H5F2NO2/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1-2H,10H2,(H,11,12)
Inchi Key MJHZRTJMDQYAHN-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-35-difluorobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-35-difluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart