APO448899930 Apollo Scientific Ltd

2,3-Difluoro-1-(trifluoromethoxy)benzene

  • MDL number MFCD18641903
  • Cas Number : 158178-35-9
  • Molecular Weight : 198.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-difluorophenyl trifluoromethyl ether
Inchi Code 1S/C7H3F5O/c8-4-2-1-3-5(6(4)9)13-7(10,11)12/h1-3H
Inchi Key IEWSBWIUOSSOJG-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-difluoro-1-trifluoromethoxybenzene

Safety Information

signalword Warning
pcodes P210;P302+P352;P303+P361+P353
MSDS https://store.apolloscientific.co.uk/product/23-difluoro-1-trifluoromethoxybenzene
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart