APO448899934 Apollo Scientific Ltd

2,5-Difluoro-1-(trifluoromethoxy)benzene

  • MDL number MFCD22580924
  • Cas Number : 1404195-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=73509

Safety Information

signalword Warning
pcodes P210;P280;P303+P361+P353
MSDS http://www.apolloscientific.co.uk/display_item.php?id=73509
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart