APO448899939 Apollo Scientific Ltd

3,5-Difluoro-1-(trifluoromethoxy)benzene

  • MDL number MFCD22581043
  • Cas Number : 1404194-50-8
  • Molecular Weight : 198.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-difluorophenyl trifluoromethyl ether
Inchi Code 1S/C7H3F5O/c8-4-1-5(9)3-6(2-4)13-7(10,11)12/h1-3H
Inchi Key LDNQHCRDKTWUGG-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-difluoro-1-trifluoromethoxybenzene

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluoro-1-trifluoromethoxybenzene
pictograms GHS02;GHS07
hstatements H225;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart