APO455823851 Apollo Scientific Ltd

2-Bromo-4,6-difluorobenzyl alcohol

  • MDL number MFCD28737914
  • Cas Number : 1807071-21-1
  • Molecular Weight : 223.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 43
Iupac Name (2-bromo-4,6-difluorophenyl)methanol
Inchi Code 1S/C7H5BrF2O/c8-6-1-4(9)2-7(10)5(6)3-11/h1-2,11H,3H2
Inchi Key QROQRUHMWGVTGW-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 43
Product Link https://store.apolloscientific.co.uk/product/2-bromo-46-difluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-46-difluorobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart