APO455824100 Apollo Scientific Ltd

2-Fluoro-3-nitroaniline

  • MDL number MFCD11046757
  • Cas Number : 21397-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-116
Storage temp. Store at 2-8°C
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-nitroaniline

Safety Information

signalword Warning
pcodes P261;P280;P312
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-nitroaniline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart