APO455824108 Apollo Scientific Ltd

5-Bromo-2-nitrobenzenesulphonyl chloride

  • MDL number MFCD12912807
  • Cas Number : 1193388-84-9
  • Molecular Weight : 300.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-88
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-nitrobenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-nitrobenzenesulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart