APO455824143 Apollo Scientific Ltd

2-Fluoro-3-formylbenzoic acid

  • MDL number MFCD20526383
  • Cas Number : 1289043-16-8
  • Molecular Weight : 168.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-3-formylbenzoic acid
Inchi Code 1S/C8H5FO3/c9-7-5(4-10)2-1-3-6(7)8(11)12/h1-4H,(H,11,12)
Inchi Key YJTYMNFESVPEKM-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-formylbenzoic-acid

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-formylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart