APO455824159 Apollo Scientific Ltd

3-Fluoro-5-(hydroxymethyl)benzoic acid

  • MDL number MFCD20659871
  • Cas Number : 816449-67-9
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-5-(hydroxymethyl)benzoic acid
Inchi Code 1S/C8H7FO3/c9-7-2-5(4-10)1-6(3-7)8(11)12/h1-3,10H,4H2,(H,11,12)
Inchi Key IMJHSBTXOXRZBD-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-hydroxymethylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-hydroxymethylbenzoic-acid
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart