APO455824240 Apollo Scientific Ltd

2-Bromo-4,6-difluorobenzaldehyde

  • MDL number MFCD18642940
  • Cas Number : 154650-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-bromo-46-difluorobenzaldehyde-tech

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-46-difluorobenzaldehyde-tech
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart