APO455825537 Apollo Scientific Ltd

5-Bromo-2-chlorophenol

  • MDL number MFCD00672940
  • Cas Number : 183802-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-58
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 234
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chlorophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chlorophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart