APO455825540 Apollo Scientific Ltd

Adamantan-2-one

  • MDL number MFCD00074737
  • Cas Number : 700-58-3
  • Molecular Weight : 150.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 257(subl.)
Synonym Tricyclo[3.3.1.1~3,7~]decan-2-one, 2-Oxotricyclo[3.3.1.1~3,7~]decane
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/adamantan-2-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/adamantan-2-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart