APO455825555 Apollo Scientific Ltd

8-Hydrazinoquinoline dihydrochloride

  • MDL number MFCD00491379
  • Cas Number : 91004-61-4
  • Molecular Weight : 232.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Hydrazino-1-azanaphthalene dihydrochloride, (Quinolin-8-yl)hydrazine dihydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-hydrazinoquinoline-dihydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/8-hydrazinoquinoline-dihydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart