APO455825568 Apollo Scientific Ltd

1-[2-(Pyridin-2-yl)ethyl]piperazine

  • MDL number MFCD01748645
  • Cas Number : 53345-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[2-(Piperazin-1-yl)ethyl]pyridine
Purity 97
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15923

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15923
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart