APO455825574 Apollo Scientific Ltd

1-(2,2-Diethoxyethyl)piperazine

  • MDL number MFCD03094589
  • Cas Number : 82516-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (Piperazin-1-yl)acetaldehyde diethyl acetal
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15931

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15931
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart