APO455825578 Apollo Scientific Ltd

2-Acetyl-3H-benzo[f]chromen-3-one

  • MDL number MFCD00445975
  • Cas Number : 727-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Acetyl-3-oxo-3H-benzo[f]chromene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15938

Safety Information

signalword Warning
pcodes P280;P301+P312;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15938
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart