APO455825579 Apollo Scientific Ltd

4-(Dimethoxymethyl)-6-hydroxy-2-thiopyrimidine

  • MDL number MFCD00662704
  • Cas Number : 90370-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-179
Synonym 4-(Dimethoxymethyl)-6-hydroxypyrimidine-2-thiol, 6-(Dimethoxymethyl)-2-sulphanylpyrimidin-4-ol
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14377

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14377
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart