APO455825589 Apollo Scientific Ltd

3-Chloro-2-methylbenzenesulphonamide

  • MDL number MFCD00234295
  • Cas Number : 3476-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-6-sulphamoyltoluene, 2-(Aminosulphonyl)-6-chlorotoluene
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15954

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=15954
Availability details for
Quantity
  • Availability
  • Add to Cart