APO455825590 Apollo Scientific Ltd

4-Bromo-3-methylbenzenesulphonamide

  • MDL number MFCD03094591
  • Cas Number : 879487-75-9
  • Molecular Weight : 250.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 148
Iupac Name 4-bromo-3-methylbenzenesulfonamide
Inchi Code 1S/C7H8BrNO2S/c1-5-4-6(12(9,10)11)2-3-7(5)8/h2-4H,1H3,(H2,9,10,11)
Inchi Key KRUSKWRPOCHSGC-UHFFFAOYSA-N

Description

Synonym 2-Bromo-5-sulphamoyltoluene
Purity 99%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 148
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-methylbenzenesulphonamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-methylbenzenesulphonamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart