APO455825596 Apollo Scientific Ltd

Ethyl (naphth-2-yl)(oxo)acetate

  • MDL number MFCD01934866
  • Cas Number : 73790-09-7
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl (naphth-2-yl)glyoxylate, 2-[Ethoxy(oxo)acetyl]naphthalene
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 138-140/0.15mm
Product Link https://store.apolloscientific.co.uk/product/ethyl-naphth-2-yloxoacetate

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-naphth-2-yloxoacetate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart