APO455825597 Apollo Scientific Ltd

4-Methyl-3-[(thien-3-yl)methyl]-5-thio-4H-1,2,4-triazole

  • MDL number MFCD00832943
  • Cas Number : 262610-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-133
Synonym 4-Methyl-5-[(thien-3-yl)methyl]-4H-1,2,4-triazole-3-thiol, 3-[(4-Methyl-5-sulphanyl-4H-1,2,4-triazol-3-yl)methyl]thiophene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15976

Safety Information

signalword Danger
pcodes P280;P210
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15976
pictograms GHS02;GHS07
hstatements H228;H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart