APO455825599 Apollo Scientific Ltd

Methyl 2,4-dioxo-4-pyridin-3-ylbutanoate

  • MDL number MFCD06245328
  • Cas Number : 23424-36-4
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-24-dioxo-4-pyridin-3-ylbutanoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-24-dioxo-4-pyridin-3-ylbutanoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart