APO455825604 Apollo Scientific Ltd

Quinoxaline-5-carboxylic acid

  • MDL number MFCD02854494
  • Cas Number : 6924-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167-172
Synonym 5-Carboxyquinoxaline, 5-Carboxy-1,4-benzodiazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/quinoxaline-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/quinoxaline-5-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart