APO455825607 Apollo Scientific Ltd

Phenyl(phenylamino)acetic acid

  • MDL number MFCD01724775
  • Cas Number : 3684-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Anilino(phenyl)acetic acid, Phenyl(phenylamino)ethanoic acid
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38781

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38781
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart