APO455825613 Apollo Scientific Ltd

2,4-Dimethylphenacyl bromide

  • MDL number MFCD00017872
  • Cas Number : 26346-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-45
Synonym 2-Bromo-1-(2,4-dimethylphenyl)ethan-1-one, 4-(Bromoacetyl)-m-xylene
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13243

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13243
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart