APO455825631 Apollo Scientific Ltd

3-Chloro-4-hydroxytetrahydrothiophene 1,1-dioxide

  • MDL number MFCD00005486
  • Cas Number : 49592-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-167
Synonym 4-Chlorotetrahydrothiophene-3-ol 1,1-dioxide, 4-Chloro-3-hydroxythiolane 1,1-dioxide
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13752

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13752
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart