APO455825633 Apollo Scientific Ltd

1-[5-(4-Chlorophenyl)-2-methylfur-3-yl]ethan-1-one

  • MDL number MFCD00832873
  • Cas Number : 43020-12-8
  • Molecular Weight : 234.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-115
Synonym 3-Acetyl-5-(4-chlorophenyl)-2-methylfuran, 1-(4-Acetyl-5-methylfur-2-yl)-4-chlorobenzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-5-4-chlorophenyl-2-methylfur-3-ylethan-1-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-5-4-chlorophenyl-2-methylfur-3-ylethan-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart