APO455825635 Apollo Scientific Ltd

4-Chloro-1H-pyrazole-3-carboxaldehyde

  • MDL number MFCD00110641
  • Cas Number : 623570-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-1H-pyrazole-5-carbaldehyde
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chloro-1h-pyrazole-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-1h-pyrazole-3-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart