APO455825636 Apollo Scientific Ltd

2-Bromo-1-(4-isobutylphenyl)propan-1-one

  • MDL number MFCD00831700
  • Cas Number : 80336-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-62
Synonym 2-Bromo-4'-(2-methylprop-1-yl)propiophenone
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13298

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13298
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart