APO455825648 Apollo Scientific Ltd

1-(Diphenylmethyl)azetidine-3-carboxylic acid

  • MDL number MFCD01566539
  • Cas Number : 36476-87-6
  • Molecular Weight : 267.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198(dec.)
Synonym 1-Benzhydrylazetidine-3-carboxylic acid, 1-Benzhydrylazetane-3-carboxylic acid
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-diphenylmethylazetidine-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-diphenylmethylazetidine-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart