APO455825650 Apollo Scientific Ltd

N-Benzylethane-1,2-diamine, N-BOC protected

  • MDL number MFCD06245501
  • Cas Number : 152193-00-5
  • Molecular Weight : 250.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl (2-aminoethyl)benzylcarbamate, N1-Benzyl-N1-(tert-butoxycarbonyl)ethane-1,2-diamine
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-benzylethane-12-diamine-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-benzylethane-12-diamine-n-boc-protected
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart