APO455825652 Apollo Scientific Ltd

3-Amino-3-methylbutanoic acid

  • MDL number MFCD06245495
  • Cas Number : 625-05-8
  • Molecular Weight : 117.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232-233
Synonym 3-Amino-3-methylbutyric acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-3-methylbutanoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-3-methylbutanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart