APO455825660 Apollo Scientific Ltd

2,2'-(Benzylimino)diethanol

  • MDL number MFCD00020587
  • Cas Number : 101-32-6
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[Benzyl(2-hydroxyethyl)amino]ethan-1-ol, N,N-Bis(2-hydroxyethyl)benzylamine, N-Benzyldiethanolamine
Purity 95
Physical Form Liquid-Viscous
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/22-benzyliminodiethanol

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/22-benzyliminodiethanol
pictograms GHS05
hstatements H318;H302
Availability details for
Quantity
  • Availability
  • Add to Cart