APO455825661 Apollo Scientific Ltd

3-(Thien-2-yl)piperazine, N1-BOC protected

  • MDL number MFCD06660372
  • Cas Number : 886771-38-6
  • Molecular Weight : 268.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(2-thienyl)-1-piperazinecarboxylate
Inchi Code 1S/C13H20N2O2S/c1-13(2,3)17-12(16)15-7-6-14-10(9-15)11-5-4-8-18-11/h4-5,8,10,14H,6-7,9H2,1-3H3
Inchi Key NSUDSXYQBVEAOK-UHFFFAOYSA-N

Description

Synonym tert-Butyl 3-(thien-2-yl)piperazine-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-(thien-2-yl)piperazine
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-thien-2-ylpiperazine-n1-boc-protected

Safety Information

signalword Warning
pcodes P280;P271
MSDS https://store.apolloscientific.co.uk/product/3-thien-2-ylpiperazine-n1-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart