APO455825663 Apollo Scientific Ltd

3-Phenylquinoxaline-5-carboxylic acid

  • MDL number MFCD06660374
  • Cas Number : 162135-93-5
  • Molecular Weight : 250.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-phenylquinoxaline-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-phenylquinoxaline-5-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart