APO455825665 Apollo Scientific Ltd

4-(Chloromethyl)-6-hydroxy-2-phenylpyrimidine

  • MDL number MFCD00113662
  • Cas Number : 35252-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-198
Synonym 6-(Chloromethyl)-2-phenylpyrimidin-4-ol, 4-(Chloromethyl)-6-hydroxy-2-phenyl-1,3-diazine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13795

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13795
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart